missing translation for 'onlineSavingsMsg'
Learn More

5-Methylbenzo[b]thiophene, 97%

Catalog No. aaa1266606
Change view
missing translation for 'orderingAttributeHoverText'
Quantity:
1 g
5 g
Catalog No. Quantity
AAA1266606 5 g
AAA1266603 1 g
2 options

Catalog No. AAA1266606

Supplier: Thermo Scientific Chemicals A1266606

Only null left

CAS: 14315-14-1 | C9H8S | 148.22 g/mol

5-Methylbenzo[b]thiophene is useful in substituted thiophene for proteomics research.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
5-Methylbenzo[b]thiophene is useful in substituted thiophene for proteomics research.

Solubility
Insoluble in water.

Notes
Store in cool dry place in tightly closed container. With good ventilation. Store away from oxidizing agent.

Chemical Identifiers

CAS 14315-14-1
Molecular Formula C9H8S
Molecular Weight (g/mol) 148.22
MDL Number MFCD00052509
InChI Key DOHZWDWNQFZIKH-UHFFFAOYSA-N
Synonym 5-methylbenzo b thiophene, benzo b thiophene, 5-methyl, 5-methylbenzothiophene, 5-methyl-benzo b thiophene, 5-methylthianaphthene, 5-methyl-benzothiophene, benzothiophene, 5-mehyl, 5-methyl benzo b thiophene
PubChem CID 84346
IUPAC Name 5-methyl-1-benzothiophene
SMILES CC1=CC=C2SC=CC2=C1

Specifications

Melting Point 32°C to 36°C
Density 1.11
Boiling Point 71°C to 73°C (0.6 mmHg)
Quantity 5 g
Beilstein 109866
Solubility Information Insoluble in water.
Formula Weight 148.23
Percent Purity 97%
Chemical Name or Material 5-Methylbenzo[b]thiophene
Hazard Category H302-H315-H319-H335
Hazard Statement GHS H Statement
H301
Toxic if swallowed.
Precautionary Statement P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c
DOTInformation Hazard Class: 9; Packaging Group: III
EINECSNumber 238-256-2
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only