missing translation for 'onlineSavingsMsg'
Learn More

5-Methylbenzimidazole 98.0+%, TCI America™

Catalog No. M13215G
Change view
Click to view available options
Quantity:
5 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
M13215G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. M13215G Supplier TCI America Supplier No. M13215G

Chemical Identifiers

CAS 614-97-1
Molecular Formula C8H8N2
Molecular Weight (g/mol) 132.166
MDL Number MFCD00010740
InChI Key RWXZXCZBMQPOBF-UHFFFAOYSA-N
Synonym 5-methylbenzimidazole, 5-methyl-1h-benzo d imidazole, 1h-benzimidazole, 5-methyl, 5-methyl-1h-benzimidazole, benzimidazole, 5-methyl, ethyl 4-cyanophenylalaninate, 1h-benzimidazole, 6-methyl, 5-methyl-1h-1,3-benzodiazole, 6-methyl-1h-1,3-benzodiazole, 5mb
PubChem CID 11979
ChEBI CHEBI:40205
IUPAC Name 6-methyl-1H-benzimidazole
SMILES CC1=CC2=C(C=C1)N=CN2

Specifications

Melting Point 112°C
Boiling Point 172°C
Quantity 5 g
Formula Weight 132.17
Percent Purity ≥98.0% (GC,T)
Physical Form Crystalline Powder
Chemical Name or Material 5-Methylbenzimidazole
RTECSNumber DD9275000
TSCA Yes