accessibility menu, dialog, popup

UserName

CH3NHN

CAS RN 614-97-1

IUPAC Name: 6-methyl-1H-1,3-benzodiazole
Synonyms: 5-methylbenzimidazole ethyl 4-cyanophenylalaninate
Molecular Weight (g/mol): 132.17
Molecular Formula: C8H8N2
InChi Key: RWXZXCZBMQPOBF-UHFFFAOYSA-N
SMILES: CC1=CC=C2N=CNC2=C1
Boiling Point 
  • (3)
  • (1)
Percent Purity 
  • (1)
  • (3)
Quantity 
  • (1)
  • (1)
  • (2)
Physical Form 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (1)
  • (3)
  • (3)
  • (1)
  • (1)
  • (3)
  • (1)
  • (1)
  • (2)
  • (1)