missing translation for 'onlineSavingsMsg'
Learn More

5-Methylbenzimidazole, 98%

CAS: 614-97-1 | C8H8N2 | 132.166 g/mol

$119.33 - $1041.53

Chemical Identifiers

CAS 614-97-1
Molecular Formula C8H8N2
Molecular Weight (g/mol) 132.166
MDL Number MFCD00010740
InChI Key RWXZXCZBMQPOBF-UHFFFAOYSA-N
Synonym 5-methylbenzimidazole, 5-methyl-1h-benzo d imidazole, 1h-benzimidazole, 5-methyl, 5-methyl-1h-benzimidazole, benzimidazole, 5-methyl, ethyl 4-cyanophenylalaninate, 1h-benzimidazole, 6-methyl, 5-methyl-1h-1,3-benzodiazole, 6-methyl-1h-1,3-benzodiazole, 5mb
PubChem CID 11979
ChEBI CHEBI:40205
IUPAC Name 6-methyl-1H-benzimidazole
SMILES CC1=CC2=C(C=C1)N=CN2
View More Specs

Products 3
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAA1595706
View Documents
Thermo Scientific Chemicals
A1595706
5 g
Each for $119.33
Only null left
 
AAA1595714
View Documents
Thermo Scientific Chemicals
A1595714
25 g
Each for $328.34
Only null left
 
AAA1595722
View Documents
Thermo Scientific Chemicals
A1595722
100 g
Each for $1,041.53
Only null left
 
Description

Description

5-Methylbenzimidazole is used as a camphor substitute. It is also used as fine chemical and medicine intermediate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
5-Methylbenzimidazole is used as a camphor substitute. It is also used as fine chemical and medicine intermediate.

Solubility
Soluble in chloroform (25mg/ml).

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Incompatible with strong oxidizing agents.
Specifications

Chemical Identifiers

614-97-1
132.166
RWXZXCZBMQPOBF-UHFFFAOYSA-N
11979
6-methyl-1H-benzimidazole
C8H8N2
MFCD00010740
5-methylbenzimidazole, 5-methyl-1h-benzo d imidazole, 1h-benzimidazole, 5-methyl, 5-methyl-1h-benzimidazole, benzimidazole, 5-methyl, ethyl 4-cyanophenylalaninate, 1h-benzimidazole, 6-methyl, 5-methyl-1h-1,3-benzodiazole, 6-methyl-1h-1,3-benzodiazole, 5mb
CHEBI:40205
CC1=CC2=C(C=C1)N=CN2

Specifications

614-97-1
169°C to 172°C (1 mmHg)
MFCD00010740
2628
Soluble in chloroform (25mg/ml).
CC1=CC2=C(C=C1)N=CN2
132.166
CHEBI:40205
98%
112°C to 117°C
C8H8N2
5 g
5-methylbenzimidazole, 5-methyl-1h-benzo d imidazole, 1h-benzimidazole, 5-methyl, 5-methyl-1h-benzimidazole, benzimidazole, 5-methyl, ethyl 4-cyanophenylalaninate, 1h-benzimidazole, 6-methyl, 5-methyl-1h-1,3-benzodiazole, 6-methyl-1h-1,3-benzodiazole, 5mb
RWXZXCZBMQPOBF-UHFFFAOYSA-N
6-methyl-1H-benzimidazole
11979
132.17
5-Methylbenzimidazole
Videos
Safety and Handling

Safety and Handling

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

P264b-P270-P301+P312-P330-P501c

H302

EINECSNumber : 210-401-4

RTECSNumber : DD9275000

TSCA : Yes

SDS
missing translation for 'documents'

missing translation for 'documents'

RUO – Research Use Only