missing translation for 'onlineSavingsMsg'
Learn More

5-Methylbenzimidazole, 98%

Catalog No. AAA1595706
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1595706 5 g
AAA1595714 25 g
AAA1595722 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAA1595706 Supplier Thermo Scientific Chemicals Supplier No. A1595706

CAS: 614-97-1 | C8H8N2 | 132.166 g/mol

5-Methylbenzimidazole is used as a camphor substitute. It is also used as fine chemical and medicine intermediate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
5-Methylbenzimidazole is used as a camphor substitute. It is also used as fine chemical and medicine intermediate.

Solubility
Soluble in chloroform (25mg/ml).

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Incompatible with strong oxidizing agents.

Chemical Identifiers

CAS 614-97-1
Molecular Formula C8H8N2
Molecular Weight (g/mol) 132.166
MDL Number MFCD00010740
InChI Key RWXZXCZBMQPOBF-UHFFFAOYSA-N
Synonym 5-methylbenzimidazole, 5-methyl-1h-benzo d imidazole, 1h-benzimidazole, 5-methyl, 5-methyl-1h-benzimidazole, benzimidazole, 5-methyl, ethyl 4-cyanophenylalaninate, 1h-benzimidazole, 6-methyl, 5-methyl-1h-1,3-benzodiazole, 6-methyl-1h-1,3-benzodiazole, 5mb
PubChem CID 11979
ChEBI CHEBI:40205
IUPAC Name 6-methyl-1H-benzimidazole
SMILES CC1=CC2=C(C=C1)N=CN2

Specifications

Melting Point 112°C to 117°C
Boiling Point 169°C to 172°C (1 mmHg)
Quantity 5 g
Beilstein 2628
Solubility Information Soluble in chloroform (25mg/ml).
Formula Weight 132.17
Percent Purity 98%
Chemical Name or Material 5-Methylbenzimidazole
Hazard Category H302
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P264b-P270-P301+P312-P330-P501c
EINECSNumber 210-401-4
RTECSNumber DD9275000
TSCA Yes

RUO – Research Use Only