missing translation for 'onlineSavingsMsg'
Learn More

5-Methyl-3-phenylisoxazole-4-carboxylic acid, 99%

CAS: 1136-45-4 | C11H9NO3 | 203.197 g/mol

Supplier:  Thermo Scientific Chemicals L0114222

 View more versions of this product

Catalog No. AAL0114222

Only null left
Explore available promotions

Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Specifications

5-Methyl-3-phenylisoxazole-4-carboxylic acid
191°C to 193°C
C11H9NO3
100 g
5-methyl-3-phenylisoxazole-4-carboxylic acid, 4-isoxazolecarboxylic acid, 5-methyl-3-phenyl, 5-methyl-3-phenyl-4-isoxazolecarboxylic acid, 3-phenyl-5-methylisoxazol-4-carbonsaeure, 3-phenyl-5-methylisoxazole-4-carboxylic acid, isoxazole-4-carboxylic acid, 5-methyl-3-phenyl, 5-methyl-3-phenyl-isoxazole-4-carboxylic acid, 5-methyl-3-phenylisoxazole-4-carboxylicacid, 3-phenyl-5-methylisoxazol-4-carbonsaeure german, 5-methyl-3-phenyl-4-isooxazolic acid
CC1=C(C(=NO1)C2=CC=CC=C2)C(=O)O
203.197
203.2
1136-45-4
Odorless
MFCD00003153
164939
PENHKTNQUJMHIR-UHFFFAOYSA-N
5-methyl-3-phenyl-1,2-oxazole-4-carboxylic acid
14343
99%

Chemical Identifiers

1136-45-4
203.197
PENHKTNQUJMHIR-UHFFFAOYSA-N
14343
CC1=C(C(=NO1)C2=CC=CC=C2)C(=O)O
C11H9NO3
MFCD00003153
5-methyl-3-phenylisoxazole-4-carboxylic acid, 4-isoxazolecarboxylic acid, 5-methyl-3-phenyl, 5-methyl-3-phenyl-4-isoxazolecarboxylic acid, 3-phenyl-5-methylisoxazol-4-carbonsaeure, 3-phenyl-5-methylisoxazole-4-carboxylic acid, isoxazole-4-carboxylic acid, 5-methyl-3-phenyl, 5-methyl-3-phenyl-isoxazole-4-carboxylic acid, 5-methyl-3-phenylisoxazole-4-carboxylicacid, 3-phenyl-5-methylisoxazol-4-carbonsaeure german, 5-methyl-3-phenyl-4-isooxazolic acid
5-methyl-3-phenyl-1,2-oxazole-4-carboxylic acid
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

GHS H Statement
H302
Harmful if swallowed.

GHS P Statement
P264-P270-P301+P312-P330-P501a
Wash thoroughly after handling.
Do not eat, drink or smoke when using this product.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell. Rinse mouth.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Warning

EINECSNumber : 214-497-9

RTECSNumber : NY2750000

TSCA : Yes

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Promotions

Promotions

RUO – Research Use Only