missing translation for 'onlineSavingsMsg'
Learn More

5-Methyl-3-phenylisoxazole-4-carboxylic acid, 99%

Catalog No. p-7043525
Change view
Click to view available options
Quantity:
25 g
100 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAL0114214 25 g
AAL0114222 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAL0114214 Supplier Thermo Scientific Chemicals Supplier No. L0114214

CAS: 1136-45-4 | C11H9NO3 | 203.197 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 1136-45-4
Molecular Formula C11H9NO3
Molecular Weight (g/mol) 203.197
MDL Number MFCD00003153
InChI Key PENHKTNQUJMHIR-UHFFFAOYSA-N
Synonym 5-methyl-3-phenylisoxazole-4-carboxylic acid, 4-isoxazolecarboxylic acid, 5-methyl-3-phenyl, 5-methyl-3-phenyl-4-isoxazolecarboxylic acid, 3-phenyl-5-methylisoxazol-4-carbonsaeure, 3-phenyl-5-methylisoxazole-4-carboxylic acid, isoxazole-4-carboxylic acid, 5-methyl-3-phenyl, 5-methyl-3-phenyl-isoxazole-4-carboxylic acid, 5-methyl-3-phenylisoxazole-4-carboxylicacid, 3-phenyl-5-methylisoxazol-4-carbonsaeure german, 5-methyl-3-phenyl-4-isooxazolic acid
PubChem CID 14343
IUPAC Name 5-methyl-3-phenyl-1,2-oxazole-4-carboxylic acid
SMILES CC1=C(C(=NO1)C2=CC=CC=C2)C(=O)O

Specifications

Melting Point 191°C to 193°C
Odor Odorless
Quantity 25 g
Beilstein 164939
Formula Weight 203.2
Percent Purity 99%
Chemical Name or Material 5-Methyl-3-phenylisoxazole-4-carboxylic acid
Hazard Category Warning
Hazard Statement GHS H Statement
H302
Harmful if swallowed.
Precautionary Statement GHS P Statement
P264-P270-P301+P312-P330-P501a
Wash thoroughly after handling.
Do not eat, drink or smoke when using this product.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell. Rinse mouth.
Dispose of contents/container in accordance with local/regional/national/international regulations.
EINECSNumber 214-497-9
RTECSNumber NY2750000
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only