missing translation for 'onlineSavingsMsg'
Learn More

5-Methyl-3-phenyl-4-isoxazolesulfonyl chloride, 97%, Thermo Scientific™

Supplier:  Thermo Scientific Chemicals CC00503DA

Quantity 1g
Packaging Amber Glass Bottle

 View more versions of this product

Catalog No. CC00503DA

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description

Description

Specifications

Chemical Identifiers

857283-56-8
257.688
GXQBTNFENYVPFM-UHFFFAOYSA-N
18525696
CC1=C(C(=NO1)C2=CC=CC=C2)S(=O)(=O)Cl
C10H8ClNO3S
MFCD06658966
5-methyl-3-phenyl-4-isoxazolesulfonyl chloride, 5-methyl-3-phenylisoxazole-4-sulphonyl chloride, 5-methyl-3-phenylisoxazole-4-sulfonyl chloride, chloro 5-methyl-3-phenylisoxazol-4-yl sulfone, 5-methyl-3-phenyl-isoxazole-4-sulfonyl chloride
5-methyl-3-phenyl-1,2-oxazole-4-sulfonyl chloride

Specifications

5-Methyl-3-phenyl-4-isoxazolesulfonyl chloride
96.5
97%
C10H8ClNO3S
MFCD06658966
GXQBTNFENYVPFM-UHFFFAOYSA-N
5-methyl-3-phenyl-1,2-oxazole-4-sulfonyl chloride
18525696
97%
857283-56-8
100.0
Amber Glass Bottle
1g
5-methyl-3-phenyl-4-isoxazolesulfonyl chloride, 5-methyl-3-phenylisoxazole-4-sulphonyl chloride, 5-methyl-3-phenylisoxazole-4-sulfonyl chloride, chloro 5-methyl-3-phenylisoxazol-4-yl sulfone, 5-methyl-3-phenyl-isoxazole-4-sulfonyl chloride
CC1=C(C(=NO1)C2=CC=CC=C2)S(=O)(=O)Cl
257.688
257.7
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

GHS H Statement:
Causes severe skin burns and eye damage.
Causes serious eye damage.
Contact with water liberates toxic gas.
Reacts violently with water.

GHS P Statement:
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.
Store in a dry place. Store in a closed container.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Danger

SDS
Documents

Documents