missing translation for 'onlineSavingsMsg'
Learn More

5-Methyl-3-phenyl-4-isoxazolesulfonyl chloride, 97%, Thermo Scientific™

Chemical Identifiers

CAS 857283-56-8
Molecular Formula C10H8ClNO3S
Molecular Weight (g/mol) 257.688
MDL Number MFCD06658966
InChI Key GXQBTNFENYVPFM-UHFFFAOYSA-N
Synonym 5-methyl-3-phenyl-4-isoxazolesulfonyl chloride, 5-methyl-3-phenylisoxazole-4-sulphonyl chloride, 5-methyl-3-phenylisoxazole-4-sulfonyl chloride, chloro 5-methyl-3-phenylisoxazol-4-yl sulfone, 5-methyl-3-phenyl-isoxazole-4-sulfonyl chloride
PubChem CID 18525696
IUPAC Name 5-methyl-3-phenyl-1,2-oxazole-4-sulfonyl chloride
SMILES CC1=C(C(=NO1)C2=CC=CC=C2)S(=O)(=O)Cl
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Packaging Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Packaging Price Quantity & Availability  
CC00503CB
View Documents
N/A 250mg Amber Glass Bottle N/A
N/A
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
CC00503DA
View Documents
N/A 1g Amber Glass Bottle N/A
N/A
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description

Description

Specifications

Chemical Identifiers

857283-56-8
257.688
GXQBTNFENYVPFM-UHFFFAOYSA-N
18525696
CC1=C(C(=NO1)C2=CC=CC=C2)S(=O)(=O)Cl
C10H8ClNO3S
MFCD06658966
5-methyl-3-phenyl-4-isoxazolesulfonyl chloride, 5-methyl-3-phenylisoxazole-4-sulphonyl chloride, 5-methyl-3-phenylisoxazole-4-sulfonyl chloride, chloro 5-methyl-3-phenylisoxazol-4-yl sulfone, 5-methyl-3-phenyl-isoxazole-4-sulfonyl chloride
5-methyl-3-phenyl-1,2-oxazole-4-sulfonyl chloride

Specifications

857283-56-8
100.0
C10H8ClNO3S
5-methyl-3-phenyl-4-isoxazolesulfonyl chloride, 5-methyl-3-phenylisoxazole-4-sulphonyl chloride, 5-methyl-3-phenylisoxazole-4-sulfonyl chloride, chloro 5-methyl-3-phenylisoxazol-4-yl sulfone, 5-methyl-3-phenyl-isoxazole-4-sulfonyl chloride
CC1=C(C(=NO1)C2=CC=CC=C2)S(=O)(=O)Cl
257.688
257.7
5-Methyl-3-phenyl-4-isoxazolesulfonyl chloride
96.5
97%
MFCD06658966
GXQBTNFENYVPFM-UHFFFAOYSA-N
5-methyl-3-phenyl-1,2-oxazole-4-sulfonyl chloride
18525696
97%
Videos
Safety and Handling

Safety and Handling

GHS H Statement:
Causes severe skin burns and eye damage.
Causes serious eye damage.
Contact with water liberates toxic gas.
Reacts violently with water.

GHS P Statement:
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.
Store in a dry place. Store in a closed container.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Danger

SDS
Documents

Documents