missing translation for 'onlineSavingsMsg'
Learn More

5-Methyl-3-phenyl-1,2,4-oxadiazole, 97%

Catalog No. AAB2014503
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAB2014503 1 g
AAB2014506 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAB2014503 Supplier Thermo Scientific Chemicals Supplier No. B2014503
Get Up to $1,800 in Chemicals with Purchase
View all promotions

CAS: 1198-98-7 | C9H8N2O | 160.18 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 1198-98-7
Molecular Formula C9H8N2O
Molecular Weight (g/mol) 160.18
MDL Number MFCD00085133
InChI Key VRRLZUXQTZOCKJ-UHFFFAOYSA-N
Synonym phenylmethyloxadiazole, 5-methyl-3-phenyl-1,2,4-oxadiazol, 1,2,4-oxadiazole, 5-methyl-3-phenyl, 1,4-oxadiazole, 5-methyl-3-phenyl, 3-phenyl-5-methyl-1,2,4-oxadiazole, 5-methyl-3-phenyl-1,2,4 oxadiazole, 1,2,4-oxadiazole, 5-methyl-3-phenyl-8ci 9ci
PubChem CID 98806
IUPAC Name 5-methyl-3-phenyl-1,2,4-oxadiazole
SMILES CC1=NC(=NO1)C1=CC=CC=C1

Specifications

Melting Point 33°C to 36°C
Quantity 1 g
Formula Weight 223.28
Percent Purity 97%
Chemical Name or Material 5-Methyl-3-phenyl-1,2,4-oxadiazole
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only