CAS RN 1198-98-7
IUPAC Name:
5-methyl-3-phenyl-1,2,4-oxadiazole
Synonyms:
5-methyl-3-phenyl-1,2,4-oxadiazole
Molecular Weight (g/mol):
160.18
Molecular Formula:
C9H8N2O
InChi Key:
VRRLZUXQTZOCKJ-UHFFFAOYSA-N
SMILES:
CC1=NC(=NO1)C1=CC=CC=C1
1
–
1
of
1
results
1
Promotions Available
5-Methyl-3-phenyl-1,2,4-oxadiazole, 97%
5-Methyl-3-phenyl-1,2,4-oxadiazole, 97%
CAS: 1198-98-7 Molecular Formula: C9H8N2O Molecular Weight (g/mol): 160.18 MDL Number: MFCD00085133 InChI Key: VRRLZUXQTZOCKJ-UHFFFAOYSA-N Synonym: phenylmethyloxadiazole,5-methyl-3-phenyl-1,2,4-oxadiazol,1,2,4-oxadiazole, 5-methyl-3-phenyl,1,4-oxadiazole, 5-methyl-3-phenyl,3-phenyl-5-methyl-1,2,4-oxadiazole,5-methyl-3-phenyl-1,2,4 oxadiazole,1,2,4-oxadiazole, 5-methyl-3-phenyl-8ci 9ci PubChem CID: 98806 IUPAC Name: 5-methyl-3-phenyl-1,2,4-oxadiazole SMILES: CC1=NC(=NO1)C1=CC=CC=C1