missing translation for 'onlineSavingsMsg'
Learn More
Learn More
5-Methyl-3-heptanone 95.0+%, TCI America™
$35.41 - $76.43
Chemical Identifiers
| CAS | 541-85-5 |
|---|---|
| Molecular Formula | C8H16O |
| Molecular Weight (g/mol) | 128.22 |
| MDL Number | MFCD00009340 |
| InChI Key | PSBKJPTZCVYXSD-UHFFFAOYNA-N |
| Synonym | 5-methyl-3-heptanone, 3-heptanone, 5-methyl, ethyl sec-amyl ketone, ethyl 2-methylbutyl ketone, 3-methyl-5-heptanone, ethyl amylketone, 2-methylbutyl ethyl ketone, 5-methyl-3-heptanon, ethyl amyl cetone, 5-methyl 3-heptanone |
| PubChem CID | 7822 |
| IUPAC Name | 5-methylheptan-3-one |
| SMILES | CCC(C)CC(=O)CC |
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|
M033525ML
|
TCI America
M033525ML |
25 mL |
Each for $35.41
|
|
|||||
|
M0335500ML
|
TCI America
M0335500ML |
500 mL |
Each for $76.43
|
|
|||||
Chemical Identifiers
| 541-85-5 | |
| 128.22 | |
| PSBKJPTZCVYXSD-UHFFFAOYNA-N | |
| 7822 | |
| CCC(C)CC(=O)CC |
| C8H16O | |
| MFCD00009340 | |
| 5-methyl-3-heptanone, 3-heptanone, 5-methyl, ethyl sec-amyl ketone, ethyl 2-methylbutyl ketone, 3-methyl-5-heptanone, ethyl amylketone, 2-methylbutyl ethyl ketone, 5-methyl-3-heptanon, ethyl amyl cetone, 5-methyl 3-heptanone | |
| 5-methylheptan-3-one |
Specifications
| 541-85-5 | |
| Yellow | |
| C8H16O | |
| 25 mL | |
| 5-methyl-3-heptanone, 3-heptanone, 5-methyl, ethyl sec-amyl ketone, ethyl 2-methylbutyl ketone, 3-methyl-5-heptanone, ethyl amylketone, 2-methylbutyl ethyl ketone, 5-methyl-3-heptanon, ethyl amyl cetone, 5-methyl 3-heptanone | |
| CCC(C)CC(=O)CC | |
| 128.22 | |
| 128.22 | |
| Liquid |
| -57°C | |
| 157°C | |
| MFCD00009340 | |
| 2271 | |
| PSBKJPTZCVYXSD-UHFFFAOYNA-N | |
| 5-methylheptan-3-one | |
| 7822 | |
| ≥95.0% (GC) | |
| 5-Methyl-3-heptanone |
Safety and Handling
EINECSNumber : (2)-0544
RTECSNumber : MJ7350000
TSCA : Yes