missing translation for 'onlineSavingsMsg'
Learn More
Learn More
5-Methyl-3-heptanone 95.0+%, TCI America™
Supplier: TCI America M033525ML
Specifications
| 5-Methyl-3-heptanone | |
| -57°C | |
| 157°C | |
| MFCD00009340 | |
| 2271 | |
| PSBKJPTZCVYXSD-UHFFFAOYNA-N | |
| 5-methylheptan-3-one | |
| 7822 | |
| ≥95.0% (GC) |
| 541-85-5 | |
| Yellow | |
| C8H16O | |
| 25 mL | |
| 5-methyl-3-heptanone, 3-heptanone, 5-methyl, ethyl sec-amyl ketone, ethyl 2-methylbutyl ketone, 3-methyl-5-heptanone, ethyl amylketone, 2-methylbutyl ethyl ketone, 5-methyl-3-heptanon, ethyl amyl cetone, 5-methyl 3-heptanone | |
| CCC(C)CC(=O)CC | |
| 128.22 | |
| 128.22 | |
| Liquid |
Chemical Identifiers
| 541-85-5 | |
| 128.22 | |
| PSBKJPTZCVYXSD-UHFFFAOYNA-N | |
| 7822 | |
| CCC(C)CC(=O)CC |
| C8H16O | |
| MFCD00009340 | |
| 5-methyl-3-heptanone, 3-heptanone, 5-methyl, ethyl sec-amyl ketone, ethyl 2-methylbutyl ketone, 3-methyl-5-heptanone, ethyl amylketone, 2-methylbutyl ethyl ketone, 5-methyl-3-heptanon, ethyl amyl cetone, 5-methyl 3-heptanone | |
| 5-methylheptan-3-one |
Safety and Handling
EINECSNumber : (2)-0544
RTECSNumber : MJ7350000
TSCA : Yes