missing translation for 'onlineSavingsMsg'
Learn More

5-Methyl-2-(trifluoromethyl)-3-furoic acid, 97%, Thermo Scientific™

Supplier:  Thermo Scientific Chemicals CC00701DA

Quantity 1g

 View more versions of this product

Catalog No. CC00701DA

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description

Description

Specifications

Chemical Identifiers

17515-74-1
194.11
NURNFMNHTHXTDS-UHFFFAOYSA-N
2782183
CC1=CC(C(O)=O)=C(O1)C(F)(F)F
C7H5F3O3
MFCD00275563
5-methyl-2-trifluoromethyl furan-3-carboxylic acid, 5-methyl-2-trifluoromethyl-3-furoic acid, 5-methyl-2-trifluoromethyl-furan-3-carboxylic acid, 2-trifluoromethyl-5-methyl-3-furancarboxylic acid, 3-furancarboxylic acid, 5-methyl-2-trifluoromethyl, 2-trifluoromethyl-5-methyl-3-furoic acid, 2-trifluoromethyl-5-methylfuran-3-carboxylic acid, 5-methyl-2-trifluoromethyl-3-furancarboxylic acid, 3-furancarboxylic acid,5-methyl-2-trifluoromethyl
5-methyl-2-(trifluoromethyl)furan-3-carboxylic acid

Specifications

5-Methyl-2-(trifluoromethyl)-3-furoic acid
97%
C7H5F3O3
1g
NURNFMNHTHXTDS-UHFFFAOYSA-N
5-methyl-2-(trifluoromethyl)furan-3-carboxylic acid
2782183
97%
17515-74-1
Amber glass bottle
MFCD00275563
5-methyl-2-trifluoromethyl furan-3-carboxylic acid, 5-methyl-2-trifluoromethyl-3-furoic acid, 5-methyl-2-trifluoromethyl-furan-3-carboxylic acid, 2-trifluoromethyl-5-methyl-3-furancarboxylic acid, 3-furancarboxylic acid, 5-methyl-2-trifluoromethyl, 2-trifluoromethyl-5-methyl-3-furoic acid, 2-trifluoromethyl-5-methylfuran-3-carboxylic acid, 5-methyl-2-trifluoromethyl-3-furancarboxylic acid, 3-furancarboxylic acid,5-methyl-2-trifluoromethyl
CC1=CC(C(O)=O)=C(O1)C(F)(F)F
194.11
194.11
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Promotions

Promotions