missing translation for 'onlineSavingsMsg'
Learn More

5-Methyl-2-(trifluoromethyl)-3-furoic acid, 97%, Thermo Scientific™

Catalog No. p-4509798
Change view
Click to view available options
Quantity:
250mg
1g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
CC00701CB 250mg
CC00701DA 1g
CC00701DE 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. CC00701CB Supplier Thermo Scientific Chemicals Supplier No. CC00701CB

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 17515-74-1
Molecular Formula C7H5F3O3
Molecular Weight (g/mol) 194.11
MDL Number MFCD00275563
InChI Key NURNFMNHTHXTDS-UHFFFAOYSA-N
Synonym 5-methyl-2-trifluoromethyl furan-3-carboxylic acid, 5-methyl-2-trifluoromethyl-3-furoic acid, 5-methyl-2-trifluoromethyl-furan-3-carboxylic acid, 2-trifluoromethyl-5-methyl-3-furancarboxylic acid, 3-furancarboxylic acid, 5-methyl-2-trifluoromethyl, 2-trifluoromethyl-5-methyl-3-furoic acid, 2-trifluoromethyl-5-methylfuran-3-carboxylic acid, 5-methyl-2-trifluoromethyl-3-furancarboxylic acid, 3-furancarboxylic acid,5-methyl-2-trifluoromethyl
PubChem CID 2782183
IUPAC Name 5-methyl-2-(trifluoromethyl)furan-3-carboxylic acid
SMILES CC1=CC(C(O)=O)=C(O1)C(F)(F)F

Specifications

Assay Percent Range 97%
Packaging Amber glass bottle
Quantity 250mg
Formula Weight 194.11
Percent Purity 97%
Chemical Name or Material 5-Methyl-2-(trifluoromethyl)-3-furoic acid