missing translation for 'onlineSavingsMsg'
Learn More
Learn More
5-Methyl-2-octanone 80.0+%, TCI America™
$146.31 - $1312.97
Chemical Identifiers
| CAS | 58654-67-4 |
|---|---|
| Molecular Formula | C9H18O |
| Molecular Weight (g/mol) | 142.24 |
| MDL Number | MFCD00059414 |
| InChI Key | UPVLGOPSYZTLGB-UHFFFAOYNA-N |
| Synonym | 5-methyl-2-octanone, 2-octanone, 5-methyl, methyl 3-methylhexyl ketone, methyloctanone, acmc-1axv1, dsstox_cid_21444, dsstox_gsid_41444, 3-01-00-02890 beilstein handbook reference |
| PubChem CID | 42802 |
| IUPAC Name | 5-methyloctan-2-one |
| SMILES | CCCC(C)CCC(C)=O |
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|
M027825ML
|
TCI America
M027825ML |
25 mL |
Each for $146.31
|
|
|||||
|
M0278500ML
|
TCI America
M0278500ML |
500 mL |
Each for $1,312.97
|
|
|||||
Chemical Identifiers
| 58654-67-4 | |
| 142.24 | |
| UPVLGOPSYZTLGB-UHFFFAOYNA-N | |
| 42802 | |
| CCCC(C)CCC(C)=O |
| C9H18O | |
| MFCD00059414 | |
| 5-methyl-2-octanone, 2-octanone, 5-methyl, methyl 3-methylhexyl ketone, methyloctanone, acmc-1axv1, dsstox_cid_21444, dsstox_gsid_41444, 3-01-00-02890 beilstein handbook reference | |
| 5-methyloctan-2-one |
Specifications
| 58654-67-4 | |
| 102°C | |
| MFCD00059414 | |
| 1224 | |
| UPVLGOPSYZTLGB-UHFFFAOYNA-N | |
| 5-methyloctan-2-one | |
| 42802 | |
| ≥80.0% (GC) | |
| 5-Methyl-2-octanone |
| Yellow | |
| C9H18O | |
| 25 mL | |
| 5-methyl-2-octanone, 2-octanone, 5-methyl, methyl 3-methylhexyl ketone, methyloctanone, acmc-1axv1, dsstox_cid_21444, dsstox_gsid_41444, 3-01-00-02890 beilstein handbook reference | |
| CCCC(C)CCC(C)=O | |
| 142.24 | |
| 142.24 | |
| Liquid |
Safety and Handling
EINECSNumber : (2)-0542
RTECSNumber : RH1535000
TSCA : Yes