missing translation for 'onlineSavingsMsg'
Learn More
Learn More
5-Methyl-2-octanone 80.0+%, TCI America™
Supplier: TCI America M027825ML
Specifications
| 5-Methyl-2-octanone | |
| Yellow | |
| C9H18O | |
| 25 mL | |
| 5-methyl-2-octanone, 2-octanone, 5-methyl, methyl 3-methylhexyl ketone, methyloctanone, acmc-1axv1, dsstox_cid_21444, dsstox_gsid_41444, 3-01-00-02890 beilstein handbook reference | |
| CCCC(C)CCC(C)=O | |
| 142.24 | |
| 142.24 | |
| Liquid |
| 58654-67-4 | |
| 102°C | |
| MFCD00059414 | |
| 1224 | |
| UPVLGOPSYZTLGB-UHFFFAOYNA-N | |
| 5-methyloctan-2-one | |
| 42802 | |
| ≥80.0% (GC) |
Chemical Identifiers
| 58654-67-4 | |
| 142.24 | |
| UPVLGOPSYZTLGB-UHFFFAOYNA-N | |
| 42802 | |
| CCCC(C)CCC(C)=O |
| C9H18O | |
| MFCD00059414 | |
| 5-methyl-2-octanone, 2-octanone, 5-methyl, methyl 3-methylhexyl ketone, methyloctanone, acmc-1axv1, dsstox_cid_21444, dsstox_gsid_41444, 3-01-00-02890 beilstein handbook reference | |
| 5-methyloctan-2-one |
Safety and Handling
EINECSNumber : (2)-0542
RTECSNumber : RH1535000
TSCA : Yes