missing translation for 'onlineSavingsMsg'
Learn More

5-Methyl-2-hexene (cis- and trans- mixture) 99.0+%, TCI America™

Catalog No. m03051ml
Change view
Click to view available options
Quantity:
1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
M03051ML 1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. M03051ML Supplier TCI America Supplier No. M03051ML
Only null left

Chemical Identifiers

CAS 3404-62-4
Molecular Formula C7H14
Molecular Weight (g/mol) 98.189
MDL Number MFCD00059239
InChI Key GHBKCPRDHLITSE-SNAWJCMRSA-N
PubChem CID 5357252
IUPAC Name (E)-5-methylhex-2-ene
SMILES CC=CCC(C)C

Specifications

Color Colorless
Quantity 1 mL
UN Number 3295
Formula Weight 98.19
Percent Purity ≥99.0% (GC)
Physical Form Liquid
Chemical Name or Material 5-Methyl-2-hexene (cis- and trans- mixture)