accessibility menu, dialog, popup

UserName

CAS RN 3404-62-4

IUPAC Name: (2E)-5-methylhex-2-ene
Synonyms: 2-hexene, 5-methyl-, (E)-
Molecular Weight (g/mol): 98.19
Molecular Formula: C7H14
InChi Key: GHBKCPRDHLITSE-SNAWJCMRSA-N
SMILES: C\C=C\CC(C)C

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results