missing translation for 'onlineSavingsMsg'
Learn More

5-Methyl-2-hexanone, 99%

Catalog No. p-164139
Change view
Click to view available options
Quantity:
1 L
2.5 L
Packaging:
Glass bottle
Catalog No. Quantity Packaging
AC149660010 1 L Glass bottle
AC149660025 2.5 L Glass bottle
2 options

Catalog No. AC149660010

Supplier: Thermo Scientific Chemicals 149660010

Only null left

CAS: 110-12-3 | C7H14O | 114.19 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 110-12-3
Molecular Formula C7H14O
Molecular Weight (g/mol) 114.19
MDL Number MFCD00008950
InChI Key FFWSICBKRCICMR-UHFFFAOYSA-N
Synonym 5-methyl-2-hexanone, 2-hexanone, 5-methyl, isoamyl methyl ketone, methyl isoamyl ketone, miak, isopentyl methyl ketone, 2-methyl-5-hexanone, ketone, methyl isoamyl, methyl isopentyl ketone, unii-6o4a4a5f28
PubChem CID 8034
IUPAC Name 5-methylhexan-2-one
SMILES CC(C)CCC(=O)C

Specifications

CAS Min % 98.5
CAS Max % 100.0
Melting Point -74.0°C
Color Colorless
Density 0.8100g/mL
Boiling Point 145.0°C
Flash Point 41°C
Infrared Spectrum Authentic
Assay Percent Range 98.5% min. (GC)
Packaging Glass bottle
Refractive Index 1.4060 to 1.4080
Linear Formula (CH3)2CHCH2CH2COCH3
Quantity 1 L
Specific Gravity 0.81
Solubility Information Solubility in water: 5.4g/L (20°C). Other solubilities: miscible with most common organic solvents
Formula Weight 114.19
Percent Purity 99%
Grade Extra Pure
Physical Form Liquid
Chemical Name or Material 5-Methyl-2-hexanone, 99%
Show More Show Less
Hazard Category GHS Signal Word: Warning
Hazard Statement GHS H Statement:
Harmful if inhaled.
Flammable liquid and vapor.
Precautionary Statement GHS P Statement:
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
EINECSNumber 203-737-8
RTECSNumber MP3850000
TSCA TSCA

RUO – Research Use Only