missing translation for 'onlineSavingsMsg'
Learn More

5-Methyl-2-heptene (cis- and trans- mixture) 95.0+%, TCI America™

Catalog No. p-7132483
Change view
Click to view available options
Quantity:
1 mL
5 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
M09921ML 1 mL
M09925ML 5 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. M09921ML Supplier TCI America Supplier No. M09921ML
Only null left

Chemical Identifiers

CAS 22487-87-2
Molecular Formula C8H16
Molecular Weight (g/mol) 112.22
MDL Number MFCD00061016
InChI Key VIHUHUGDEZCPDK-UHFFFAOYNA-N
PubChem CID 5364849
IUPAC Name 5-methylhept-2-ene
SMILES CCC(C)CC=CC

Specifications

Color Colorless
Quantity 1 mL
UN Number 3295
Formula Weight 112.22
Percent Purity ≥95.0% (GC)
Physical Form Liquid
Chemical Name or Material 5-Methyl-2-heptene (cis- and trans- mixture)