accessibility menu, dialog, popup

UserName

CH3CH3H3C(E/Z)

CAS RN 22487-87-2

IUPAC Name: 5-methylhept-2-ene
Synonyms: 5-methylhept-2-ene
Molecular Weight (g/mol): 112.22
Molecular Formula: C8H16
InChi Key: VIHUHUGDEZCPDK-UHFFFAOYNA-N
SMILES: CCC(C)CC=CC
Percent Purity 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results