Learn More
5-Methyl-2-furaldehyde, 98%
CAS: 620-02-0 | C6H6O2 | 110.112 g/mol
Supplier: Thermo Scientific Chemicals A1326422
Description
5-Methyl-2-furaldehyde is used as a perfuming agent. It is used a smoke with essence.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications5-Methyl-2-furaldehyde is used as a perfuming agent. It is used a smoke with essence.
Solubility
Soluble in alcohol and water.
Notes
Store at room temperature. Incompatible with oxidizing agents. Air sensitive.
Specifications
| 5-Methyl-2-furaldehyde | |
| 1.106 | |
| 72°C (161°F) | |
| 1.531 | |
| 100 g | |
| Air Sensitive | |
| Soluble in alcohol and water. | |
| CC1=CC=C(O1)C=O | |
| 110.112 | |
| CHEBI:2091 | |
| 98% |
| 620-02-0 | |
| 186°C to 188°C | |
| C6H6O2 | |
| MFCD00003232 | |
| 106895 | |
| 5-methylfurfural, 5-methyl-2-furaldehyde, 5-methyl furfural, 5-methyl-2-furfural, 2-furancarboxaldehyde, 5-methyl, 5-methylfuran-2-al, 5-methyl-2-furancarboxaldehyde, 5-methylfurfuraldehyde, 2-furaldehyde, 5-methyl, 2-formyl-5-methylfuran | |
| OUDFNZMQXZILJD-UHFFFAOYSA-N | |
| 5-methylfuran-2-carbaldehyde | |
| 12097 | |
| 110.11 |
Chemical Identifiers
| 620-02-0 | |
| 110.112 | |
| OUDFNZMQXZILJD-UHFFFAOYSA-N | |
| 12097 | |
| 5-methylfuran-2-carbaldehyde |
| C6H6O2 | |
| MFCD00003232 | |
| 5-methylfurfural, 5-methyl-2-furaldehyde, 5-methyl furfural, 5-methyl-2-furfural, 2-furancarboxaldehyde, 5-methyl, 5-methylfuran-2-al, 5-methyl-2-furancarboxaldehyde, 5-methylfurfuraldehyde, 2-furaldehyde, 5-methyl, 2-formyl-5-methylfuran | |
| CHEBI:2091 | |
| CC1=CC=C(O1)C=O |
Safety and Handling
GHS H Statement
H315-H319-H335-H227
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Combustible liquid.
P210-P235-P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P370+P378q-P501c
H227-H315-H319
EINECSNumber : 210-622-6
RTECSNumber : LT7032500
TSCA : Yes
Recommended Storage : Ambient temperatures; Store under Argon
RUO – Research Use Only