missing translation for 'onlineSavingsMsg'
Learn More

5-Methyl-2-furaldehyde, 98%

Catalog No. p-7021075
Change view
Click to view available options
Quantity:
25 g
100 g
500 g
Catalog No. Quantity
AAA1326414 25 g
AAA1326422 100 g
AAA1326436 500 g
3 options

Catalog No. AAA1326414

Supplier: Thermo Scientific Chemicals A1326414

Only null left

CAS: 620-02-0 | C6H6O2 | 110.112 g/mol

5-Methyl-2-furaldehyde is used as a perfuming agent. It is used a smoke with essence.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
5-Methyl-2-furaldehyde is used as a perfuming agent. It is used a smoke with essence.

Solubility
Soluble in alcohol and water.

Notes
Store at room temperature. Incompatible with oxidizing agents. Air sensitive.

Chemical Identifiers

CAS 620-02-0
Molecular Formula C6H6O2
Molecular Weight (g/mol) 110.112
MDL Number MFCD00003232
InChI Key OUDFNZMQXZILJD-UHFFFAOYSA-N
Synonym 5-methylfurfural, 5-methyl-2-furaldehyde, 5-methyl furfural, 5-methyl-2-furfural, 2-furancarboxaldehyde, 5-methyl, 5-methylfuran-2-al, 5-methyl-2-furancarboxaldehyde, 5-methylfurfuraldehyde, 2-furaldehyde, 5-methyl, 2-formyl-5-methylfuran
PubChem CID 12097
ChEBI CHEBI:2091
IUPAC Name 5-methylfuran-2-carbaldehyde
SMILES CC1=CC=C(O1)C=O

Specifications

Density 1.106
Boiling Point 186°C to 188°C
Flash Point 72°C (161°F)
Refractive Index 1.531
Quantity 25 g
Beilstein 106895
Sensitivity Air Sensitive
Solubility Information Soluble in alcohol and water.
Formula Weight 110.11
Percent Purity 98%
Chemical Name or Material 5-Methyl-2-furaldehyde
Show More Show Less
Hazard Category H227-H315-H319
Hazard Statement GHS H Statement
H315-H319-H335-H227
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Combustible liquid.
Precautionary Statement P210-P235-P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P370+P378q-P501c
EINECSNumber 210-622-6
RTECSNumber LT7032500
TSCA Yes
Recommended Storage Ambient temperatures; Store under Argon

RUO – Research Use Only