missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
5-Methyl-2-(2H-1,2,3-triazol-2-yl)benzoic Acid 98.0+%, TCI America™
$56.41 - $222.30
Chemical Identifiers
CAS | 956317-36-5 |
---|---|
Molecular Formula | C10H9N3O2 |
Molecular Weight (g/mol) | 203.201 |
MDL Number | MFCD18382679 |
InChI Key | SRBAGFIYKNQXDV-UHFFFAOYSA-N |
PubChem CID | 53260165 |
IUPAC Name | 5-methyl-2-(triazol-2-yl)benzoic acid |
SMILES | CC1=CC(=C(C=C1)N2N=CC=N2)C(=O)O |
Chemical Identifiers
956317-36-5 | |
203.201 | |
SRBAGFIYKNQXDV-UHFFFAOYSA-N | |
5-methyl-2-(triazol-2-yl)benzoic acid |
C10H9N3O2 | |
MFCD18382679 | |
53260165 | |
CC1=CC(=C(C=C1)N2N=CC=N2)C(=O)O |
Specifications
956317-36-5 | |
White-Yellow | |
MFCD18382679 | |
SRBAGFIYKNQXDV-UHFFFAOYSA-N | |
5-methyl-2-(triazol-2-yl)benzoic acid | |
53260165 | |
≥98.0% (HPLC,T) | |
5-Methyl-2-(2H-1,2,3-triazol-2-yl)benzoic Acid |
175°C | |
C10H9N3O2 | |
1 g | |
CC1=CC(=C(C=C1)N2N=CC=N2)C(=O)O | |
203.201 | |
203.20 | |
Crystalline Powder |
Safety and Handling
TSCA : No