missing translation for 'onlineSavingsMsg'
Learn More

5-Methyl-2-(2,2,2-trimethylacetamido)pyridine, 98%, Thermo Scientific Chemicals

Catalog No. p-7057177
Change view
Click to view available options
Quantity:
250 mg
1 g
5 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH66596MD 250 mg
AAH6659603 1 g
AAH6659606 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAH66596MD Supplier Thermo Scientific Chemicals Supplier No. H66596MD
Only null left

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 86847-78-1
Molecular Formula C11H16N2O
Molecular Weight (g/mol) 192.262
MDL Number MFCD03374646
InChI Key XUCQZJBNEVIURX-UHFFFAOYSA-N
Synonym n-5-methylpyridin-2-yl pivalamide, 2,2-dimethyl-n-5-methylpyridin-2-yl propanamide, 2,2-dimethyl-n-5-methyl-pyridin-2-yl-propionamide, 6-pivaloylamino 3-picoline, n-5-methyl-2-pyridinyl pivalamide, 2,2-dimethyl-n-5-methyl-2-pyridinyl propanamide, propanamide, 2,2-dimethyl-n-5-methyl-2-pyridinyl
PubChem CID 4182924
IUPAC Name 2,2-dimethyl-N-(5-methylpyridin-2-yl)propanamide
SMILES CC1=CN=C(C=C1)NC(=O)C(C)(C)C

Specifications

Quantity 250 mg
Beilstein 4179181
Formula Weight 192.26
Percent Purity 98%
Physical Form Solid
Chemical Name or Material 5-Methyl-2-(2,2,2-trimethylacetamido)pyridine
Hazard Category H315-H319-H335
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only