missing translation for 'onlineSavingsMsg'
Learn More

5-Methyl-1-phenylpyrazole-4-carboxylic Acid 98.0+%, TCI America™

Catalog No. p-7133373
Change view
Click to view available options
Quantity:
1 g
5 g
Catalog No. Quantity
M26921G 1 g
M26925G 5 g
2 options

Catalog No. M26921G

Supplier: TCI America M26921G

Only null left

Chemical Identifiers

CAS 91138-00-0
Molecular Formula C11H10N2O2
Molecular Weight (g/mol) 202.213
MDL Number MFCD00067831
InChI Key USSMIQWDLWJQDQ-UHFFFAOYSA-N
Synonym 5-methyl-1-phenyl-1h-pyrazole-4-carboxylic acid, 3-methyl-2-phenylpyrazole-4-carboxylic acid, 3-methyl-2-phenyl-4-pyrazolecarboxylic acid, akos pao-0331, acmc-20am2c, rarechem al be 1317, maybridge1_002413, buttpark 33\08-16, cbi-bb zero/006129, 5-methyl-1-phenyl-1h-pyrazole-4-carboxylicacid
PubChem CID 145221
IUPAC Name 5-methyl-1-phenylpyrazole-4-carboxylic acid
SMILES CC1=C(C=NN1C2=CC=CC=C2)C(=O)O

Specifications

Melting Point 169°C
Color White-Yellow
Quantity 1 g
Formula Weight 202.21
Percent Purity ≥98.0% (GC,T)
Physical Form Crystalline Powder
Chemical Name or Material 5-Methyl-1-phenylpyrazole-4-carboxylic Acid