missing translation for 'onlineSavingsMsg'
Learn More

5-Methyl-1-hexyn-3-ol 98.0+%, TCI America™

Catalog No. M09615ML
Change view
Click to view available options
Quantity:
5 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
M09615ML 5 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. M09615ML Supplier TCI America Supplier No. M09615ML

Chemical Identifiers

CAS 61996-79-0
Molecular Formula C7H12O
Molecular Weight (g/mol) 112.172
MDL Number MFCD00041606
InChI Key NTNUBJHPRAMQPC-UHFFFAOYSA-N
Synonym 5-methyl-1-hexyn-3-ol, acmc-209myn, 5-methyl-hex-1-yn-3-ol, 1-hexyn-3-ol,5-methyl, 1-hexyn-3-ol, 5-methyl
PubChem CID 143856
IUPAC Name 5-methylhex-1-yn-3-ol
SMILES CC(C)CC(C#C)O

Specifications

Color Yellow
Boiling Point 156°C
Quantity 5 mL
UN Number 1987
Formula Weight 112.17
Percent Purity ≥98.0% (GC)
Physical Form Liquid
Chemical Name or Material 5-Methyl-1-hexyn-3-ol