accessibility menu, dialog, popup

UserName

CAS RN 61996-79-0

IUPAC Name: 5-methylhex-1-yn-3-ol
Synonyms: 5-methylhex-1-yn-3-ol
Molecular Weight (g/mol): 112.17
Molecular Formula: C7H12O
InChi Key: NTNUBJHPRAMQPC-UHFFFAOYNA-N
SMILES: CC(C)CC(O)C#C
Percent Purity 
  • (1)
  • (3)
Quantity 
  • (1)
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (1)
  • (3)
  • (1)
  • (3)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)