missing translation for 'onlineSavingsMsg'
Learn More

5-Methyl-1-hexyn-3-ol, 97%

Catalog No. p-7035763
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH5349003 1 g
AAH5349006 5 g
AAH5349014 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAH5349003 Supplier Thermo Scientific Chemicals Supplier No. H5349003

CAS: 61996-79-0 | C7H12O | 112.172 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 61996-79-0
Molecular Formula C7H12O
Molecular Weight (g/mol) 112.172
MDL Number MFCD00041606
InChI Key NTNUBJHPRAMQPC-UHFFFAOYSA-N
Synonym 5-methyl-1-hexyn-3-ol, acmc-209myn, 5-methyl-hex-1-yn-3-ol, 1-hexyn-3-ol,5-methyl, 1-hexyn-3-ol, 5-methyl
PubChem CID 143856
IUPAC Name 5-methylhex-1-yn-3-ol
SMILES CC(C)CC(C#C)O

Specifications

Density 0.86
Flash Point 53°C (127°F)
Refractive Index 1.436
Quantity 1 g
UN Number UN1987
Beilstein 1741083
Formula Weight 112.17
Percent Purity 97%
Chemical Name or Material 5-Methyl-1-hexyn-3-ol
Hazard Category H226-H302
Hazard Statement GHS H Statement
H301-H226
Toxic if swallowed.
Flammable liquid and vapor.
Precautionary Statement P210-P233-P240-P241-P242-P243-P264b-P270-P280-P301+P312-P303+P361+P353-P330-P370+P378q-P501c
DOTInformation Transport Hazard Class: 3; Packing Group: III; Proper Shipping Name: ALCOHOLS, N.O.S.
TSCA No
Recommended Storage Keep cold

RUO – Research Use Only