missing translation for 'onlineSavingsMsg'
Learn More
Learn More
5-Methyl-1,3,4-thiadiazole-2-thiol 98.0+%, TCI America™
$57.79 - $57.79
Chemical Identifiers
| CAS | 29490-19-5 |
|---|---|
| Molecular Formula | C3H4N2S2 |
| Molecular Weight (g/mol) | 132.20 |
| MDL Number | MFCD00038349 |
| InChI Key | FPVUWZFFEGYCGB-UHFFFAOYSA-N |
| Synonym | 2-mercapto-5-methyl-1,3,4-thiadiazole, 5-methyl-1,3,4-thiadiazole-2-thiol, 1,3,4-thiadiazole-2 3h-thione, 5-methyl, 5-methyl-1,3,4-thiadiazole-2 3h-thione, 5-methyl-1,3,4-thiadiazol-2-thiol, 2-methyl-5-mercapto-1,3,4-thiadiazole, d2-1,3,4-thiadiazoline-5-thione, 2-methyl, cefazolin impurity e, pubchem9932, acmc-1cbwn |
| PubChem CID | 1810203 |
| IUPAC Name | 5-methyl-2,3-dihydro-1,3,4-thiadiazole-2-thione |
| SMILES | CC1=NNC(=S)S1 |
Chemical Identifiers
| 29490-19-5 | |
| 132.20 | |
| FPVUWZFFEGYCGB-UHFFFAOYSA-N | |
| 1810203 | |
| CC1=NNC(=S)S1 |
| C3H4N2S2 | |
| MFCD00038349 | |
| 2-mercapto-5-methyl-1,3,4-thiadiazole, 5-methyl-1,3,4-thiadiazole-2-thiol, 1,3,4-thiadiazole-2 3h-thione, 5-methyl, 5-methyl-1,3,4-thiadiazole-2 3h-thione, 5-methyl-1,3,4-thiadiazol-2-thiol, 2-methyl-5-mercapto-1,3,4-thiadiazole, d2-1,3,4-thiadiazoline-5-thione, 2-methyl, cefazolin impurity e, pubchem9932, acmc-1cbwn | |
| 5-methyl-2,3-dihydro-1,3,4-thiadiazole-2-thione |
Specifications
| 29490-19-5 | |
| White-Yellow | |
| MFCD00038349 | |
| 2-mercapto-5-methyl-1,3,4-thiadiazole, 5-methyl-1,3,4-thiadiazole-2-thiol, 1,3,4-thiadiazole-2 3h-thione, 5-methyl, 5-methyl-1,3,4-thiadiazole-2 3h-thione, 5-methyl-1,3,4-thiadiazol-2-thiol, 2-methyl-5-mercapto-1,3,4-thiadiazole, d2-1,3,4-thiadiazoline-5-thione, 2-methyl, cefazolin impurity e, pubchem9932, acmc-1cbwn | |
| CC1=NNC(=S)S1 | |
| 132.20 | |
| 132.20 | |
| Crystalline Powder |
| 186°C | |
| C3H4N2S2 | |
| 25 g | |
| FPVUWZFFEGYCGB-UHFFFAOYSA-N | |
| 5-methyl-2,3-dihydro-1,3,4-thiadiazole-2-thione | |
| 1810203 | |
| ≥98.0% (T) | |
| 5-Methyl-1,3,4-thiadiazole-2-thiol |
Safety and Handling
EINECSNumber : (5)-0569
TSCA : No