CAS RN 29490-19-5
IUPAC Name:
5-methyl-2,3-dihydro-1,3,4-thiadiazole-2-thione
Synonyms:
5-methyl-3H-1,3,4-thiadiazole-2-thione
Molecular Weight (g/mol):
132.2
Molecular Formula:
C3H4N2S2
InChi Key:
FPVUWZFFEGYCGB-UHFFFAOYSA-N
SMILES:
CC1=NNC(=S)S1
1
–
1
of
1
results
1
5-Methyl-1,3,4-thiadiazole-2-thiol 98.0+%, TCI America™
CAS: 29490-19-5 Molecular Formula: C3H4N2S2 Molecular Weight (g/mol): 132.20 MDL Number: MFCD00038349 InChI Key: FPVUWZFFEGYCGB-UHFFFAOYSA-N Synonym: 2-mercapto-5-methyl-1,3,4-thiadiazole,5-methyl-1,3,4-thiadiazole-2-thiol,1,3,4-thiadiazole-2 3h-thione, 5-methyl,5-methyl-1,3,4-thiadiazole-2 3h-thione,5-methyl-1,3,4-thiadiazol-2-thiol,2-methyl-5-mercapto-1,3,4-thiadiazole,d2-1,3,4-thiadiazoline-5-thione, 2-methyl,cefazolin impurity e,pubchem9932,acmc-1cbwn PubChem CID: 1810203 IUPAC Name: 5-methyl-2,3-dihydro-1,3,4-thiadiazole-2-thione SMILES: CC1=NNC(=S)S1