missing translation for 'onlineSavingsMsg'
Learn More

5-Methyl-1,3,4-thiadiazole-2-thiol 98.0+%, TCI America™

Catalog No. M097625G
Change view
Click to view available options
Quantity:
25 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
M097625G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. M097625G Supplier TCI America Supplier No. M097625G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 29490-19-5
Molecular Formula C3H4N2S2
Molecular Weight (g/mol) 132.20
MDL Number MFCD00038349
InChI Key FPVUWZFFEGYCGB-UHFFFAOYSA-N
Synonym 2-mercapto-5-methyl-1,3,4-thiadiazole, 5-methyl-1,3,4-thiadiazole-2-thiol, 1,3,4-thiadiazole-2 3h-thione, 5-methyl, 5-methyl-1,3,4-thiadiazole-2 3h-thione, 5-methyl-1,3,4-thiadiazol-2-thiol, 2-methyl-5-mercapto-1,3,4-thiadiazole, d2-1,3,4-thiadiazoline-5-thione, 2-methyl, cefazolin impurity e, pubchem9932, acmc-1cbwn
PubChem CID 1810203
IUPAC Name 5-methyl-2,3-dihydro-1,3,4-thiadiazole-2-thione
SMILES CC1=NNC(=S)S1

Specifications

Melting Point 186°C
Color White-Yellow
Quantity 25 g
Formula Weight 132.20
Percent Purity ≥98.0% (T)
Physical Form Crystalline Powder
Chemical Name or Material 5-Methyl-1,3,4-thiadiazole-2-thiol
EINECSNumber (5)-0569
TSCA No