missing translation for 'onlineSavingsMsg'
Learn More

5-Methoxytryptamine, TCI America™

Catalog No. p-7132010
Change view
Click to view available options
Quantity:
100 mg
1 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
M0131100MG 100 mg
M01311G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. M0131100MG Supplier TCI America Supplier No. M0131100MG
Only null left

Chemical Identifiers

CAS 608-07-1
Molecular Formula C11H14N2O
Molecular Weight (g/mol) 190.246
MDL Number MFCD00005662
InChI Key JTEJPPKMYBDEMY-UHFFFAOYSA-N
Synonym 5-methoxytryptamine, 2-5-methoxy-1h-indol-3-yl ethanamine, mexamine, methoxytryptamine, 3-2-aminoethyl-5-methoxyindole, 5-mot, mexamine base, 5mot, o-methylserotonin, 1h-indole-3-ethanamine, 5-methoxy
PubChem CID 1833
ChEBI CHEBI:2089
IUPAC Name 2-(5-methoxy-1H-indol-3-yl)ethanamine
SMILES COC1=CC2=C(C=C1)NC=C2CCN

Specifications

Melting Point 121°C
Quantity 100 mg
Formula Weight 190.25
Physical Form Crystalline Powder
Chemical Name or Material 5-Methoxytryptamine
RTECSNumber NL4059000
TSCA No