accessibility menu, dialog, popup

UserName

CH3OHNH2N

CAS RN 608-07-1

IUPAC Name: 2-(5-methoxy-1H-indol-3-yl)ethan-1-amine
Synonyms: 5 methoxytryptamine 5MOT mexamine O-methylserotonin 5-MeOT
Molecular Weight (g/mol): 190.25
Molecular Formula: C11H14N2O
InChi Key: JTEJPPKMYBDEMY-UHFFFAOYSA-N
SMILES: COC1=CC=C2NC=C(CCN)C2=C1
Quantity 
  • (2)
  • (1)
  • (1)
Molecular Weight (g/mol) 
  • (1)
  • (2)
Physical Form 
  • (2)
Grade 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (1)
  • (2)
  • (1)
  • (2)
  • (1)
  • (1)
  • (1)
  • (2)
  • (2)
  • (1)