missing translation for 'onlineSavingsMsg'
Learn More

5-Methoxytryptamine, 97%

Catalog No. ac398580010
Change view
Click to view available options
Quantity:
1 g
Packaging:
Glass bottle
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC398580010 1 g Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AC398580010 Supplier Thermo Scientific Chemicals Supplier No. 398580010
Only null left

CAS: 608-07-1 | C11H14N2O | 190.24 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 608-07-1
Molecular Formula C11H14N2O
Molecular Weight (g/mol) 190.24
InChI Key JTEJPPKMYBDEMY-UHFFFAOYSA-N
Synonym 5-methoxytryptamine, 2-5-methoxy-1h-indol-3-yl ethanamine, mexamine, methoxytryptamine, 3-2-aminoethyl-5-methoxyindole, 5-mot, mexamine base, 5mot, o-methylserotonin, 1h-indole-3-ethanamine, 5-methoxy
PubChem CID 1833
ChEBI CHEBI:2089
IUPAC Name 2-(5-methoxy-1H-indol-3-yl)ethanamine
SMILES COC1=CC2=C(C=C1)NC=C2CCN

Specifications

Melting Point 120.0°C
Assay Percent Range 96% min. (GC)
Packaging Glass bottle
Quantity 1 g
Merck Index 15,6076
Formula Weight 190.24
Percent Purity 97%
Chemical Name or Material 5-Methoxytryptamine
Hazard Category GHS Signal Word: Danger
Hazard Statement GHS H Statement
Causes severe skin burns and eye damage.
Harmful if swallowed.
Precautionary Statement GHS P Statement
IF SWALLOWED: rinse mouth.
Do NOT induce vomiting.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if present and easy to do.
Continue rinsi
EINECSNumber 210-153-7

RUO – Research Use Only