missing translation for 'onlineSavingsMsg'
Learn More

5-Methoxyresorcinol 95.0+%, TCI America™

Catalog No. M14131G
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
M14131G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. M14131G Supplier TCI America Supplier No. M14131G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 2174-64-3
Molecular Formula C7H8O3
Molecular Weight (g/mol) 140.138
MDL Number MFCD00002285
InChI Key HDVRLUFGYQYLFJ-UHFFFAOYSA-N
Synonym 5-methoxyresorcinol, flamenol, 3,5-dihydroxyanisole, phloroglucinol monomethyl ether, 1,3-benzenediol, 5-methoxy, 5-methoxy-benzene-1,3-diol, unii-6201e0jif3, dsstox_cid_26525, dsstox_rid_81690, dsstox_gsid_46525
PubChem CID 71648
IUPAC Name 5-methoxybenzene-1,3-diol
SMILES COC1=CC(=CC(=C1)O)O

Specifications

Color White-Yellow
Boiling Point 213°C
Quantity 1 g
Formula Weight 140.14
Percent Purity ≥95.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 5-Methoxyresorcinol