missing translation for 'onlineSavingsMsg'
Learn More

5-Methoxyresorcinol, 95%

CAS: 2174-64-3 | C7H8O3 | 140.138 g/mol

Supplier:  Thermo Scientific Chemicals H6199603

 View more versions of this product

Catalog No. AAH6199603


Only null left
Explore available promotions

Description

Description

5-Methoxyresorcinol was used in synthesis of isorobustin, substituted linear and angular benzofurocoumarins.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
5-Methoxyresorcinol was used in synthesis of isorobustin, substituted linear and angular benzofurocoumarins.

Solubility
Soluble in water.

Notes
Keep container tightly closed. Store away from oxidizing agents.
Specifications

Specifications

5-Methoxyresorcinol
78°C to 80°C
188°C to 189°C (12 mmHg)
MFCD00002285
1423578
Soluble in water.
COC1=CC(=CC(=C1)O)O
140.138
140.14
Solid
2174-64-3
Yellow
C7H8O3
1 g
5-methoxyresorcinol, flamenol, 3,5-dihydroxyanisole, phloroglucinol monomethyl ether, 1,3-benzenediol, 5-methoxy, 5-methoxy-benzene-1,3-diol, unii-6201e0jif3, dsstox_cid_26525, dsstox_rid_81690, dsstox_gsid_46525
HDVRLUFGYQYLFJ-UHFFFAOYSA-N
5-methoxybenzene-1,3-diol
71648
95%

Chemical Identifiers

2174-64-3
140.138
HDVRLUFGYQYLFJ-UHFFFAOYSA-N
71648
COC1=CC(=CC(=C1)O)O
C7H8O3
MFCD00002285
5-methoxyresorcinol, flamenol, 3,5-dihydroxyanisole, phloroglucinol monomethyl ether, 1,3-benzenediol, 5-methoxy, 5-methoxy-benzene-1,3-diol, unii-6201e0jif3, dsstox_cid_26525, dsstox_rid_81690, dsstox_gsid_46525
5-methoxybenzene-1,3-diol
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

GHS H Statement
H302-H315-H319-H335
Harmful if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c

H302-H315-H319-H335

EINECSNumber : 218-532-9

TSCA : No

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Promotions

Promotions

RUO – Research Use Only