missing translation for 'onlineSavingsMsg'
Learn More

5-Methoxyresorcinol, 95%

Catalog No. AAH61996MD
Change view
Click to view available options
Quantity:
250 mg
1 g
5 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH61996MD 250 mg
AAH6199603 1 g
AAH6199606 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAH61996MD Supplier Thermo Scientific Chemicals Supplier No. H61996MD
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 2174-64-3 | C7H8O3 | 140.138 g/mol

5-Methoxyresorcinol was used in synthesis of isorobustin, substituted linear and angular benzofurocoumarins.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
5-Methoxyresorcinol was used in synthesis of isorobustin, substituted linear and angular benzofurocoumarins.

Solubility
Soluble in water.

Notes
Keep container tightly closed. Store away from oxidizing agents.

Chemical Identifiers

CAS 2174-64-3
Molecular Formula C7H8O3
Molecular Weight (g/mol) 140.138
MDL Number MFCD00002285
InChI Key HDVRLUFGYQYLFJ-UHFFFAOYSA-N
Synonym 5-methoxyresorcinol, flamenol, 3,5-dihydroxyanisole, phloroglucinol monomethyl ether, 1,3-benzenediol, 5-methoxy, 5-methoxy-benzene-1,3-diol, unii-6201e0jif3, dsstox_cid_26525, dsstox_rid_81690, dsstox_gsid_46525
PubChem CID 71648
IUPAC Name 5-methoxybenzene-1,3-diol
SMILES COC1=CC(=CC(=C1)O)O

Specifications

Melting Point 78°C to 80°C
Color Yellow
Boiling Point 188°C to 189°C (12 mmHg)
Quantity 250 mg
Beilstein 1423578
Solubility Information Soluble in water.
Formula Weight 140.14
Percent Purity 95%
Physical Form Solid
Chemical Name or Material 5-Methoxyresorcinol
Hazard Category H302-H315-H319-H335
Hazard Statement GHS H Statement
H302-H315-H319-H335
Harmful if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c
EINECSNumber 218-532-9
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only