missing translation for 'onlineSavingsMsg'
Learn More

5-Methoxyindole-3-carboxylic Acid Hydrate 98.0+%, TCI America™

Catalog No. p-7133221
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
M24381G 1 g
M24385G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. M24381G Supplier TCI America Supplier No. M24381G
Only null left

Chemical Identifiers

CAS 10242-01-0
Molecular Formula C10H9NO3
Molecular Weight (g/mol) 191.186
MDL Number MFCD03265451
InChI Key RVVSEZGJCOAUED-UHFFFAOYSA-N
Synonym 5-methoxyindole-3-carboxylic acid, 5-methoxy-3-indolecarboxylic acid, 5-methoxy-3-carboxyindole, 1h-indole-3-carboxylic acid, 5-methoxy, pubchem7984, zlchem 1276, 1h-indole-3-carboxylicacid, 5-methoxy, 5-methoxyindole-3-carboxylic acid;, 5-methoxyindole-3-carboxylic acid 5-methoxy-1h-indazole-3-carboxylic acid
PubChem CID 259191
IUPAC Name 5-methoxy-1H-indole-3-carboxylic acid
SMILES COC1=CC2=C(C=C1)NC=C2C(=O)O

Specifications

Melting Point 173°C
Quantity 1 g
Formula Weight 191.19
Percent Purity ≥98.0% (HPLC,T)
Physical Form Crystalline Powder
Chemical Name or Material 5-Methoxyindole-3-carboxylic Acid Hydrate