missing translation for 'onlineSavingsMsg'
Learn More
Learn More
5-Methoxyindole-3-carboxylic Acid Hydrate 98.0+%, TCI America™
Specifications
Specifications
| Melting Point | 173°C |
| Quantity | 5 g |
| Formula Weight | 191.19 |
| Percent Purity | ≥98.0% (HPLC,T) |
| Physical Form | Crystalline Powder |
| Chemical Name or Material | 5-Methoxyindole-3-carboxylic Acid Hydrate |
Safety and Handling
| TSCA | No |