missing translation for 'onlineSavingsMsg'
Learn More

5-Methoxyindole-3-carboxaldehyde 95.0+%, TCI America™

Catalog No. M12685G
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
M12685G 5 g
M12681G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. M12685G Supplier TCI America Supplier No. M12685G

Chemical Identifiers

CAS 10601-19-1
Molecular Formula C10H9NO2
Molecular Weight (g/mol) 175.19
MDL Number MFCD00005623
InChI Key TUWARWGEOHQXCO-UHFFFAOYSA-N
Synonym 5-methoxyindole-3-carboxaldehyde, 3-formyl-5-methoxyindole, 5-methoxyindole-3-carboxyaldehyde, 5-methoxyindole-3-carbaldehyde, 5-methoxyindole-3-aldehyde, 5-methoxy-3-formylindole, 1h-indole-3-carboxaldehyde, 5-methoxy, 5-methoxy-indol-3-aldehyde, unii-63c50zw2d0, 5-methoxy-1h-indole-3-carboxaldehyde
PubChem CID 82758
IUPAC Name 5-methoxy-1H-indole-3-carbaldehyde
SMILES COC1=CC=C2NC=C(C=O)C2=C1

Specifications

Melting Point 181°C
Quantity 5 g
Formula Weight 175.19
Percent Purity ≥95.0% (T)
Physical Form Crystalline Powder
Chemical Name or Material 5-Methoxyindole-3-carboxaldehyde