accessibility menu, dialog, popup

UserName

CH3OHNO

CAS RN 10601-19-1

IUPAC Name: 5-methoxy-1H-indole-3-carbaldehyde
Synonyms: 5-methoxy-1H-indole-3-carbaldehyde
Molecular Weight (g/mol): 175.19
Molecular Formula: C10H9NO2
InChi Key: TUWARWGEOHQXCO-UHFFFAOYSA-N
SMILES: COC1=CC=C2NC=C(C=O)C2=C1
Percent Purity 
  • (2)
  • (2)
Quantity 
  • (2)
  • (2)
Physical Form 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)