missing translation for 'onlineSavingsMsg'
Learn More

5-Methoxyindole-3-carboxaldehyde, 99%

Catalog No. AAL1907203
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAL1907203 1 g
AAL1907206 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAL1907203 Supplier Thermo Scientific Chemicals Supplier No. L1907203
Get Up to $1,800 in Chemicals with Purchase
View all promotions

CAS: 10601-19-1 | C10H9NO2 | 175.19 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 10601-19-1
Molecular Formula C10H9NO2
Molecular Weight (g/mol) 175.19
MDL Number MFCD00005623
InChI Key TUWARWGEOHQXCO-UHFFFAOYSA-N
Synonym 5-methoxyindole-3-carboxaldehyde, 3-formyl-5-methoxyindole, 5-methoxyindole-3-carboxyaldehyde, 5-methoxyindole-3-carbaldehyde, 5-methoxyindole-3-aldehyde, 5-methoxy-3-formylindole, 1h-indole-3-carboxaldehyde, 5-methoxy, 5-methoxy-indol-3-aldehyde, unii-63c50zw2d0, 5-methoxy-1h-indole-3-carboxaldehyde
PubChem CID 82758
IUPAC Name 5-methoxy-1H-indole-3-carbaldehyde
SMILES COC1=CC=C2NC=C(C=O)C2=C1

Specifications

Melting Point 179°C to 183°C
Quantity 1 g
Beilstein 132769
Sensitivity Air Sensitive
Formula Weight 175.19
Percent Purity 99%
Chemical Name or Material 5-Methoxyindole-3-carboxaldehyde
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
EINECSNumber 234-220-5
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only