missing translation for 'onlineSavingsMsg'
Learn More

5-Methoxyindole-3-acetic Acid 98.0+%, TCI America™

Catalog No. M13031G
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
M13031G 1 g
M13035G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. M13031G Supplier TCI America Supplier No. M13031G

Chemical Identifiers

CAS 3471-31-6
Molecular Formula C11H11NO3
Molecular Weight (g/mol) 205.213
MDL Number MFCD00005638
InChI Key COCNDHOPIHDTHK-UHFFFAOYSA-N
Synonym 5-methoxyindole-3-acetic acid, 2-5-methoxy-1h-indol-3-yl acetic acid, 5-methoxyindoleacetic acid, methoxyindoleacetic acid, 5-methoxy-3-indoleacetic acid, 5-methoxyindol-3-ylacetic acid, 5-methoxy-1h-indol-3-yl acetic acid, 5-methoxyindole-3-aceticacid, 5-methoxyindoleacetate, 1h-indole-3-acetic acid, 5-methoxy
PubChem CID 18986
ChEBI CHEBI:28281
IUPAC Name 2-(5-methoxy-1H-indol-3-yl)acetic acid
SMILES COC1=CC2=C(C=C1)NC=C2CC(=O)O

Specifications

Melting Point 148°C
Quantity 1 g
Formula Weight 205.21
Percent Purity ≥98.0% (HPLC,T)
Physical Form Crystalline Powder
Chemical Name or Material 5-Methoxyindole-3-acetic Acid
RTECSNumber NL3660500
TSCA No