accessibility menu, dialog, popup

UserName

CH3OHNOO

CAS RN 3471-31-6

IUPAC Name: 2-(5-methoxy-1H-indol-3-yl)acetate
Synonyms: 5-methoxyindoleacetate
Molecular Weight (g/mol): 204.21
Molecular Formula: C11H10NO3
InChi Key: COCNDHOPIHDTHK-UHFFFAOYSA-M
SMILES: COC1=CC=C2NC=C(CC([O-])=O)C2=C1
Percent Purity 
  • (2)
  • (2)
Quantity 
  • (2)
  • (1)
  • (1)
Physical Form 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (1)
  • (1)
  • (2)