missing translation for 'onlineSavingsMsg'
Learn More

5-Methoxyindole-3-acetic acid, 98+%

Catalog No. AAL19073MD
Change view
Click to view available options
Quantity:
250 mg
1 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAL19073MD 250 mg
AAL1907303 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAL19073MD Supplier Thermo Scientific Chemicals Supplier No. L19073MD
Get Up to $1,800 in Chemicals with Purchase
View all promotions

CAS: 3471-31-6 | C11H11NO3 | 205.213 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 3471-31-6
Molecular Formula C11H11NO3
Molecular Weight (g/mol) 205.213
MDL Number MFCD00005638
InChI Key COCNDHOPIHDTHK-UHFFFAOYSA-N
Synonym 5-methoxyindole-3-acetic acid, 2-5-methoxy-1h-indol-3-yl acetic acid, 5-methoxyindoleacetic acid, methoxyindoleacetic acid, 5-methoxy-3-indoleacetic acid, 5-methoxyindol-3-ylacetic acid, 5-methoxy-1h-indol-3-yl acetic acid, 5-methoxyindole-3-aceticacid, 5-methoxyindoleacetate, 1h-indole-3-acetic acid, 5-methoxy
PubChem CID 18986
ChEBI CHEBI:28281
IUPAC Name 2-(5-methoxy-1H-indol-3-yl)acetic acid
SMILES COC1=CC2=C(C=C1)NC=C2CC(=O)O

Specifications

Melting Point 145°C to 148°C (decomposition)
Quantity 250 mg
Beilstein 187161
Formula Weight 205.22
Percent Purity ≥98%
Chemical Name or Material 5-Methoxyindole-3-acetic acid
EINECSNumber 222-438-3
RTECSNumber NL3660500
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only