missing translation for 'onlineSavingsMsg'
Learn More

5-Methoxyflavone 98.0+%, TCI America™

Catalog No. M23651G
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
M23651G 1 g
M23655G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. M23651G Supplier TCI America Supplier No. M23651G
Only null left

Chemical Identifiers

CAS 42079-78-7
Molecular Formula C16H12O3
Molecular Weight (g/mol) 252.269
MDL Number MFCD00016942
InChI Key XRQSPUXANRGDAV-UHFFFAOYSA-N
Synonym 5-methoxyflavone, 5-methoxy-2-phenyl-4h-chromen-4-one, 5-methoxy-2-phenyl-4-benzopyrone, chembl16685, 4h-1-benzopyran-4-one, 5-methoxy-2-phenyl, 5-methoxy-2-phenyl-chromen-4-one, acmc-20am2h, d0v5hf, bidd:er0473
PubChem CID 94525
IUPAC Name 5-methoxy-2-phenylchromen-4-one
SMILES COC1=CC=CC2=C1C(=O)C=C(O2)C3=CC=CC=C3

Specifications

Melting Point 132°C
Color White-Yellow
Quantity 1 g
Formula Weight 252.27
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 5-Methoxyflavone