missing translation for 'onlineSavingsMsg'
Learn More

5-Methoxy-2-nitrotoluene 98.0+%, TCI America™

Supplier:  TCI America M136925G

Catalog No. M136925G


Only null left
Explore available promotions

Description

Description

Specifications

Specifications

5-Methoxy-2-nitrotoluene
51°C
C8H9NO3
25 g
RTZOGYCMIMOVHU-UHFFFAOYSA-N
4-methoxy-2-methyl-1-nitrobenzene
79330
≥98.0% (GC)
5367-32-8
135°C
MFCD00007167
3-methyl-4-nitroanisole, 5-methoxy-2-nitrotoluene, benzene, 4-methoxy-2-methyl-1-nitro, 1-methoxy-3-methyl-4-nitrobenzene, 4-methoxy-2-methyl-1-nitro-benzene, anisole, 3-methyl-4-nitro, 3-methoxy-6-nitrotoluene, 2-nitro-5-methoxy toluene, 2-nitro-5-methoxymethylbenzene, pubchem4149
CC1=C(C=CC(=C1)OC)[N+](=O)[O-]
167.164
167.16
Crystalline Powder

Chemical Identifiers

5367-32-8
167.164
RTZOGYCMIMOVHU-UHFFFAOYSA-N
79330
CC1=C(C=CC(=C1)OC)[N+](=O)[O-]
C8H9NO3
MFCD00007167
3-methyl-4-nitroanisole, 5-methoxy-2-nitrotoluene, benzene, 4-methoxy-2-methyl-1-nitro, 1-methoxy-3-methyl-4-nitrobenzene, 4-methoxy-2-methyl-1-nitro-benzene, anisole, 3-methyl-4-nitro, 3-methoxy-6-nitrotoluene, 2-nitro-5-methoxy toluene, 2-nitro-5-methoxymethylbenzene, pubchem4149
4-methoxy-2-methyl-1-nitrobenzene
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

EINECSNumber : (3)-0799

TSCA : No

SDS
Documents

Documents

Promotions

Promotions