missing translation for 'onlineSavingsMsg'
Learn More

5-Methoxy-2-nitrophenol, 98%

Catalog No. p-3778991
Change view
Click to view available options
Quantity:
1 g
5 g
Packaging:
Glass bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC335440010 1 g Glass bottle
AC335440050 5 g Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AC335440010 Supplier Thermo Scientific Chemicals Supplier No. 335440010

CAS: 704-14-3 | C7H7NO4 | 169.14 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 704-14-3
Molecular Formula C7H7NO4
Molecular Weight (g/mol) 169.14
MDL Number MFCD00100932
InChI Key NRTULWPODYLFOJ-UHFFFAOYSA-N
PubChem CID 219635
IUPAC Name 5-methoxy-2-nitrophenol
SMILES COC1=CC=C(C(O)=C1)[N+]([O-])=O

Specifications

CAS Min % 97.5
CAS Max % 100.0
Melting Point 90°C to 94°C
Color Yellow
Infrared Spectrum Authentic
Assay Percent Range 97.5% min. (GC)
Packaging Glass bottle
Quantity 1 g
Synonym 2-hydroxy-4-methoxynitrobenzene, phenol, 5-methoxy-2-nitro, 2-nitro-5-methoxyphenol, 3-hydroxy-4-nitroanisole, 5-methoxy-2-nitro-phenol, phenol,5-methoxy-2-nitro, pubchem22230, 3-methoxy-6-nitrophenol, 2-hydroxy4-methoxynitrobenzene, 5-methoxy-2-nitrophenol
Solubility Information Solubility in water: .. Other solubilities: soluble in acetone,chloroform,ethyl alcohol
Formula Weight 169.14
Percent Purity 98%
Physical Form Crystals or Powder
Chemical Name or Material 5-Methoxy-2-nitrophenol
Show More Show Less
Hazard Category GHS Signal Word: Danger
Hazard Statement GHS H Statement
Causes skin irritation.
Causes serious eye damage.
May cause respiratory irritation.
Precautionary Statement GHS P Statement
IF ON SKIN: Wash with plenty of soap and water.
If skin irritation occurs: Get medical advice/attention.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if present and easy to do.

RUO – Research Use Only